Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139284.2 | NP_644813.1 | 4 - 5 | 984 | 78 |
XM_011526595.1 | XP_011524897.1 | 5 - 6 | 465 | 78 | |
XM_017026428.1 | XP_016881917.1 | 4 - 5 | 313 | 78 | |
XM_017026429.1 | XP_016881918.1 | 5 - 6 | 385 | 78 | |
XM_017026430.1 | XP_016881919.1 | 3 - 4 | 167 | 78 | |
GenBank mRNA | AF467954.1 | - | 4 - 5 | 676 | 78 |
AJ487519.1 | - | 4 - 5 | 909 | 78 | |
AJ487959.1 | - | 4 - 5 | 417 | 78 | |
AL832968.1 | - | 2 - 3 | 125 | 78 | |
AY358121.1 | - | 4 - 5 | 579 | 78 | |
BC029943.1 | - | 5 - 6 | 449 | 78 | |
BC087848.1 | - | 4 - 5 | 561 | 78 | |
BC136694.1 | - | 4 - 5 | 459 | 78 | |
BC136697.1 | - | 4 - 5 | 459 | 78 |