Assay Design



Gene Transcripts

Gene Symbol
UMOD
Entrez Gene ID
7369
Gene Name
uromodulin
Gene Aliases
ADMCKD2, FJHN, HNFJ, HNFJ1, MCKD2, THGP, THP
Chromosome Location
Chr.16: 20333051 - 20356301 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008389.2 NP_001008390.1 3 - 4 1092 104
NM_001278614.1 NP_001265543.1 4 - 5 1254 104
NM_003361.3 NP_003352.2 3 - 4 1155 104
XM_011545934.1 XP_011544236.1 3 - 4 1092 104
XM_011545935.1 XP_011544237.1 4 - 5 1151 104
XM_011545936.1 XP_011544238.1 4 - 5 1088 104
XM_011545937.1 XP_011544239.1 4 - 5 978 104
XM_011545938.1 XP_011544240.1 3 - 4 1010 104
XM_011545940.1 XP_011544242.1 3 - 4 1028 104
XM_017023641.1 XP_016879130.1 3 - 4 1092 104
GenBank mRNA AK091961.1 - 4 - 5 1086 104
AK096043.1 - 4 - 5 735 104
AK127643.1 - 4 - 5 588 104
AK127648.1 - 4 - 5 851 104
BC035975.1 - 4 - 5 841 104
M15881.1 - 3 - 4 1031 104
M17778.1 - 2 - 3 968 104

View Full Product Details