Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_054113.3 | NP_473454.1 | 3 - 4 | 249 | 73 |
GenBank mRNA | AB050868.1 | - | 3 - 4 | 199 | 73 |
BC069524.1 | - | 3 - 4 | 180 | 73 | |
BC112200.1 | - | 3 - 4 | 249 | 73 | |
BC113591.1 | - | 3 - 4 | 249 | 73 |