Assay Design



Gene Transcripts

Gene Symbol
CCDC90B
Entrez Gene ID
60492
Gene Name
coiled-coil domain containing 90B
Gene Aliases
MDS011, MDS025
Chromosome Location
Chr.11: 83259093 - 83286407 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286116.1 NP_001273045.1 7 - 8 805 150
NM_001286117.1 NP_001273046.1 7 - 8 1352 150
NM_001286118.1 NP_001273047.1 6 - 7 928 150
NM_001286119.1 NP_001273048.1 7 - 8 1013 150
NM_001286120.1 NP_001273049.1 5 - 6 808 150
NM_021825.4 NP_068597.2 7 - 8 1032 150
XM_005274154.2 XP_005274211.1 6 - 7 1187 150
XM_005274156.3 XP_005274213.1 7 - 8 1019 150
XM_005274159.3 XP_005274216.1 8 - 9 698 150
XM_017018103.1 XP_016873592.1 6 - 7 630 150
XM_017018104.1 XP_016873593.1 8 - 9 1123 150
GenBank mRNA AF182412.1 - 7 - 8 602 150
AF182424.1 - 7 - 8 602 150
AF271782.1 - 7 - 8 959 150
AK055972.1 - 7 - 8 1349 150
AK225726.1 - 7 - 8 613 150
AK304769.1 - 7 - 8 765 150
AK315787.1 - 7 - 8 993 150
AL136791.1 - 7 - 8 752 150
AV756714.1 - 6 - 7 568 150
BC014573.1 - 7 - 8 642 150
BC020783.1 - 7 - 8 603 150
BC067764.1 - 7 - 8 923 150
BC107755.1 - 7 - 8 773 150

View Full Product Details