Assay Design
Gene Transcripts
- Gene Symbol
- RAP2B
- Entrez Gene ID
-
5912
- Gene Name
- RAP2B, member of RAS oncogene family
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 153162212 - 153170624 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_002886.3 |
NP_002877.2 |
1 - 1 |
3918 |
133 |
View Full Product Details