Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199198.2 | NP_001186127.1 | 16 - 17 | 1889 | 113 |
NM_018309.4 | NP_060779.2 | 15 - 16 | 1844 | 113 | |
XM_011512974.2 | XP_011511276.1 | 16 - 17 | 1752 | 113 | |
XM_017006840.1 | XP_016862329.1 | 15 - 16 | 1550 | 113 | |
XM_017006841.1 | XP_016862330.1 | 16 - 17 | 1720 | 113 | |
GenBank mRNA | AB449910.1 | - | 15 - 16 | 1642 | 113 |
AK001750.1 | - | 12 - 13 | 1330 | 113 | |
AK001908.1 | - | 15 - 16 | 1649 | 113 | |
AK092926.1 | - | 1 - 2 | 325 | 113 | |
AL713684.1 | - | 8 - 9 | 770 | 113 | |
AY740521.1 | - | 5 - 6 | 550 | 113 | |
BC010221.1 | - | 6 - 7 | 587 | 113 | |
BC020955.1 | - | 16 - 17 | 1683 | 113 | |
BX648032.1 | - | 15 - 16 | 1636 | 113 |