Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199198.2 | NP_001186127.1 | 4 - 5 | 685 | 125 |
NM_018309.4 | NP_060779.2 | 4 - 5 | 685 | 125 | |
XM_011512974.2 | XP_011511276.1 | 3 - 4 | 430 | 125 | |
XM_017006840.1 | XP_016862329.1 | 4 - 5 | 517 | 125 | |
XM_017006841.1 | XP_016862330.1 | 4 - 5 | 502 | 125 | |
GenBank mRNA | AK001750.1 | - | 1 - 2 | 171 | 125 |
BC020955.1 | - | 4 - 5 | 479 | 125 | |
BX648032.1 | - | 4 - 5 | 477 | 125 |