Assay Design



Gene Transcripts

Gene Symbol
ADAM32
Entrez Gene ID
203102
Gene Name
ADAM metallopeptidase domain 32
Gene Aliases
-
Chromosome Location
Chr.8: 39107529 - 39284917 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313994.1 NP_001300923.1 14 - 15 1754 79
NM_145004.6 NP_659441.3 17 - 18 2152 79
XM_006716298.2 XP_006716361.1 17 - 18 1946 79
XM_006716299.1 XP_006716362.1 15 - 16 1628 79
XM_011544433.1 XP_011542735.1 17 - 18 1946 79
XM_011544434.1 XP_011542736.1 15 - 16 1727 79
XM_017013183.1 XP_016868672.1 16 - 17 1877 79
XM_017013184.1 XP_016868673.1 17 - 18 1946 79
XM_017013185.1 XP_016868674.1 16 - 17 1952 79
XM_017013186.1 XP_016868675.1 13 - 14 1556 79
XM_017013187.1 XP_016868676.1 9 - 10 1092 79
XM_017013188.1 XP_016868677.1 8 - 9 1061 79
GenBank mRNA AK122608.1 - 15 - 16 1507 79
AK129810.1 - 3 - 4 1054 79
AK131038.1 - 9 - 10 1121 79
AK300148.1 - 14 - 15 1712 79
AK313864.1 - 17 - 18 1973 79
AY172952.1 - 15 - 16 1756 79
AY358739.1 - 17 - 18 2032 79
BC026085.2 - 17 - 18 1936 79
BC026169.2 - 17 - 18 1963 79
BC028702.2 - 17 - 18 1946 79
BC030014.2 - 17 - 18 1939 79
BC030698.2 - 17 - 18 1954 79
BC034975.2 - 17 - 18 1947 79

View Full Product Details