Assay Design



Gene Transcripts

Gene Symbol
CHDH
Entrez Gene ID
55349
Gene Name
choline dehydrogenase
Gene Aliases
-
Chromosome Location
Chr.3: 53813130 - 53846393 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018397.4 NP_060867.2 7 - 8 1700 141
XM_006713250.3 XP_006713313.1 7 - 8 1482 141
XM_006713251.3 XP_006713314.1 7 - 8 1659 141
XM_011533938.2 XP_011532240.1 7 - 8 1516 141
XM_011533939.2 XP_011532241.1 7 - 8 2355 141
XM_017006797.1 XP_016862286.1 7 - 8 1513 141
XM_017006798.1 XP_016862287.1 7 - 8 1505 141
GenBank mRNA AJ272267.1 - 5 - 6 924 141
AK297575.1 - 3 - 4 383 141
BC034502.1 - 7 - 8 1458 141

View Full Product Details