Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100588.1 | NP_001094058.1 | 20 - 21 | 3624 | 82 |
GenBank mRNA | AF255304.1 | - | 8 - 9 | 1068 | 82 |
AK027042.1 | - | 11 - 12 | 1861 | 82 | |
AL833177.1 | - | 21 - 22 | 3481 | 82 |