Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100588.1 | NP_001094058.1 | 2 - 3 | 477 | 104 |
NM_018835.2 | NP_061323.2 | 2 - 3 | 477 | 104 | |
GenBank mRNA | AF255303.1 | - | 1 - 2 | 837 | 104 |
AL833177.1 | - | 2 - 3 | 416 | 104 | |
BC011688.2 | - | 1 - 2 | 114 | 104 | |
BC044642.1 | - | 2 - 3 | 409 | 104 | |
BU783286.1 | - | 2 - 3 | 483 | 104 |