Assay Design



Gene Transcripts

Gene Symbol
RC3H2
Entrez Gene ID
54542
Gene Name
ring finger and CCCH-type domains 2
Gene Aliases
MNAB, RNF164
Chromosome Location
Chr.9: 122849453 - 122905283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100588.1 NP_001094058.1 9 - 10 1563 97
NM_018835.2 NP_061323.2 9 - 10 1563 97
GenBank mRNA AF255303.1 - 8 - 9 1923 97
AK000308.1 - 7 - 8 997 97
AK226010.1 - 7 - 8 997 97
AL834431.1 - 3 - 4 314 97

View Full Product Details