Assay Design
Gene Transcripts
- Gene Symbol
- CNOT8
- Entrez Gene ID
-
9337
- Gene Name
- CCR4-NOT transcription complex subunit 8
- Gene Aliases
-
CAF1, CALIF, Caf1b, POP2, hCAF1
- Chromosome Location
-
Chr.5: 154857952 - 154876792 on Build GRCh38
View Full Product Details