Assay Design



Gene Transcripts

Gene Symbol
SPATA20
Entrez Gene ID
64847
Gene Name
spermatogenesis associated 20
Gene Aliases
HEL-S-98, SSP411, Tisp78
Chromosome Location
Chr.17: 50547089 - 50555852 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258372.1 NP_001245301.1 2 - 3 370 78
NM_001258373.1 NP_001245302.1 3 - 4 473 78
NM_022827.3 NP_073738.2 3 - 4 418 78
GenBank mRNA AK025000.1 - 2 - 3 267 78
AK092731.1 - 3 - 4 78
AK125807.1 - 5 - 6 78
AK225189.1 - 2 - 3 267 78
AK291805.1 - 3 - 4 78
AK302850.1 - 2 - 3 317 78
AK308616.1 - 3 - 4 78
AK310416.1 - 2 - 3 262 78
BC025255.1 - 3 - 4 306 78
BC065526.1 - 3 - 4 78
BC111029.1 - 2 - 3 275 78
BM787432.1 - 2 - 3 333 78
CR936769.1 - 2 - 3 78
DA295930.1 - 3 - 4 433 78
DA489234.1 - 3 - 4 78
DQ238597.1 - 2 - 3 336 78
GQ472220.1 - 2 - 3 275 78

View Full Product Details