Assay Design



Gene Transcripts

Gene Symbol
SPATA20
Entrez Gene ID
64847
Gene Name
spermatogenesis associated 20
Gene Aliases
HEL-S-98, SSP411, Tisp78
Chromosome Location
Chr.17: 50547089 - 50555852 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258372.1 NP_001245301.1 7 - 8 1061 81
NM_001258373.1 NP_001245302.1 8 - 9 1164 81
NM_022827.3 NP_073738.2 8 - 9 1109 81
GenBank mRNA AK025000.1 - 7 - 8 958 81
AK025622.1 - 3 - 4 350 81
AK092731.1 - 8 - 9 1094 81
AK225189.1 - 7 - 8 958 81
AK291805.1 - 8 - 9 1052 81
AK310416.1 - 7 - 8 953 81
BC025255.1 - 8 - 9 997 81
BC065526.1 - 8 - 9 1051 81
BC111029.1 - 7 - 8 966 81
CR936769.1 - 7 - 8 979 81
DQ238597.1 - 7 - 8 1027 81
GQ472220.1 - 7 - 8 966 81

View Full Product Details