Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145354.1 | NP_001138826.1 | 9 - 10 | 977 | 72 |
NM_001321316.1 | NP_001308245.1 | 8 - 9 | 908 | 72 | |
NM_013255.4 | NP_037387.2 | 8 - 9 | 905 | 72 | |
XM_006715993.2 | XP_006716056.1 | 8 - 9 | 905 | 72 | |
XM_011516224.2 | XP_011514526.1 | 8 - 9 | 905 | 72 | |
GenBank mRNA | AK002024.1 | - | 8 - 9 | 847 | 72 |
AK294384.1 | - | 9 - 10 | 978 | 72 | |
AK310452.1 | - | 8 - 9 | 858 | 72 | |
BC002834.2 | - | 3 - 4 | 283 | 72 | |
BC067825.1 | - | 8 - 9 | 862 | 72 |