Assay Design



Gene Transcripts

Gene Symbol
FBXO22
Entrez Gene ID
26263
Gene Name
F-box protein 22
Gene Aliases
FBX22, FISTC1
Chromosome Location
Chr.15: 75903859 - 75937513 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012170.3 NP_036302.1 1 - 2 245 56
NM_147188.2 NP_671717.1 1 - 2 245 56
XM_011521447.2 XP_011519749.1 1 - 2 165 56
GenBank mRNA AK024048.1 - 1 - 2 226 56
AK223500.1 - 1 - 2 202 56
AK307084.1 - 1 - 2 228 56
AY005144.1 - 1 - 2 140 56
BC020204.1 - 1 - 2 245 56
BC032540.1 - 1 - 2 163 56
BC041691.1 - 1 - 2 212 56
BG328756.1 - 1 - 2 245 56

View Full Product Details