Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021185.4 | NP_067008.3 | 9 - 10 | 1229 | 59 |
XM_005259114.4 | XP_005259171.1 | 9 - 10 | 1325 | 59 | |
XM_005259115.4 | XP_005259172.1 | 9 - 10 | 1248 | 59 | |
XM_006723308.3 | XP_006723371.1 | 9 - 10 | 1845 | 59 | |
XM_006723310.2 | XP_006723373.1 | 9 - 10 | 1229 | 59 | |
XM_006723311.2 | XP_006723374.1 | 9 - 10 | 1250 | 59 | |
XM_011527174.2 | XP_011525476.1 | 9 - 10 | 1239 | 59 | |
XM_011527175.2 | XP_011525477.1 | 9 - 10 | 1145 | 59 | |
XM_011527176.2 | XP_011525478.1 | 9 - 10 | 1229 | 59 | |
XM_011527177.2 | XP_011525479.1 | 9 - 10 | 1109 | 59 | |
XM_011527178.2 | XP_011525480.1 | 10 - 11 | 1591 | 59 | |
XM_011527179.1 | XP_011525481.1 | 9 - 10 | 1229 | 59 | |
XM_011527182.2 | XP_011525484.1 | 8 - 9 | 973 | 59 | |
XM_017027053.1 | XP_016882542.1 | 9 - 10 | 1229 | 59 | |
XM_017027054.1 | XP_016882543.1 | 7 - 8 | 842 | 59 | |
GenBank mRNA | AK128220.1 | - | 9 - 10 | 1012 | 59 |
AK310460.1 | - | 5 - 6 | 559 | 59 | |
AL133023.1 | - | 9 - 10 | 1845 | 59 | |
BC094785.1 | - | 9 - 10 | 1149 | 59 | |
BC109035.1 | - | 5 - 6 | 628 | 59 |