Assay Design



Gene Transcripts

Gene Symbol
MIR22HG
Entrez Gene ID
84981
Gene Name
MIR22 host gene
Gene Aliases
C17orf91
Chromosome Location
Chr.17: 1711504 - 1716272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_028502.1 2 - 2 1288 64
GenBank mRNA AF070569.1 - 1 - 1 857 64
BX648321.1 - 2 - 2 1225 64

View Full Product Details