Assay Design



Gene Transcripts

Gene Symbol
FAM189A2
Entrez Gene ID
9413
Gene Name
family with sequence similarity 189 member A2
Gene Aliases
C9orf61, X123
Chromosome Location
Chr.9: 69324572 - 69392461 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127608.1 NP_001121080.1 4 - 5 362 65
NM_004816.3 NP_004807.3 4 - 5 678 65
XM_005252307.4 XP_005252364.1 4 - 5 1239 65
XM_017015324.1 XP_016870813.1 3 - 4 423 65
XM_017015325.1 XP_016870814.1 4 - 5 362 65
GenBank mRNA AK056152.1 - 2 - 3 306 65
AK126127.1 - 4 - 5 678 65
BC021685.1 - 1 - 2 91 65
BC113683.1 - 4 - 5 321 65
BC113685.1 - 4 - 5 321 65
BX641153.1 - 1 - 2 5531 65
DA824091.1 - 4 - 5 362 65

View Full Product Details