Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012324.4 | NP_036456.1 | 3 - 4 | 582 | 89 |
XM_011530679.2 | XP_011528981.1 | 3 - 4 | 471 | 89 | |
XM_011530680.2 | XP_011528982.1 | 3 - 4 | 471 | 89 | |
XM_011530681.2 | XP_011528983.1 | 3 - 4 | 471 | 89 | |
GenBank mRNA | AF136382.1 | - | 3 - 4 | 565 | 89 |
AF218778.1 | - | 1 - 2 | 367 | 89 | |
AK294265.1 | - | 3 - 4 | 472 | 89 | |
BC047527.1 | - | 3 - 4 | 495 | 89 |