Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012324.4 | NP_036456.1 | 6 - 7 | 1971 | 86 |
XM_011530679.2 | XP_011528981.1 | 6 - 7 | 1863 | 86 | |
XM_011530680.2 | XP_011528982.1 | 6 - 7 | 1749 | 86 | |
XM_011530681.2 | XP_011528983.1 | 6 - 7 | 1728 | 86 | |
GenBank mRNA | AF136382.1 | - | 6 - 7 | 1954 | 86 |
AF218778.1 | - | 4 - 5 | 1756 | 86 | |
AK293580.1 | - | 4 - 5 | 342 | 86 | |
AK294265.1 | - | 7 - 8 | 1030 | 86 | |
AL021708.1 | - | 2 - 3 | 974 | 86 | |
BC009940.2 | - | 5 - 6 | 721 | 86 | |
BC047527.1 | - | 6 - 7 | 1884 | 86 | |
U79261.1 | - | 1 - 2 | 45 | 86 |