Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012324.4 | NP_036456.1 | 7 - 8 | 2143 | 109 |
XM_011530679.2 | XP_011528981.1 | 7 - 8 | 2035 | 109 | |
XM_011530680.2 | XP_011528982.1 | 7 - 8 | 1921 | 109 | |
XM_011530681.2 | XP_011528983.1 | 7 - 8 | 1900 | 109 | |
GenBank mRNA | AF136382.1 | - | 7 - 8 | 2126 | 109 |
AF218778.1 | - | 5 - 6 | 1928 | 109 | |
AK293580.1 | - | 5 - 6 | 514 | 109 | |
AK294265.1 | - | 8 - 9 | 1202 | 109 | |
AL021708.1 | - | 3 - 4 | 1146 | 109 | |
BC009940.2 | - | 6 - 7 | 893 | 109 | |
BC047527.1 | - | 7 - 8 | 2056 | 109 | |
U79261.1 | - | 2 - 3 | 217 | 109 |