Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012324.4 | NP_036456.1 | 1 - 2 | 201 | 83 |
XM_011530679.2 | XP_011528981.1 | 1 - 2 | 90 | 83 | |
XM_011530680.2 | XP_011528982.1 | 1 - 2 | 90 | 83 | |
XM_011530681.2 | XP_011528983.1 | 1 - 2 | 90 | 83 | |
GenBank mRNA | AF136382.1 | - | 1 - 2 | 184 | 83 |
AK293580.1 | - | 1 - 2 | 114 | 83 | |
AK294265.1 | - | 1 - 2 | 91 | 83 | |
BC009940.2 | - | 1 - 2 | 94 | 83 | |
BC047527.1 | - | 1 - 2 | 114 | 83 | |
DB483256.1 | - | 1 - 2 | 203 | 83 |