Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003803.3 | NP_003794.3 | 20 - 21 | 3454 | 86 |
NM_019856.1 | NP_062830.1 | 19 - 20 | 3166 | 86 | |
XM_017026062.1 | XP_016881551.1 | 20 - 21 | 3181 | 86 | |
GenBank mRNA | AJ621424.1 | - | 20 - 21 | 3454 | 86 |
AK125322.1 | - | 21 - 22 | 3454 | 86 | |
BC116183.1 | - | 19 - 20 | 2907 | 86 | |
X69090.1 | - | 17 - 18 | 2538 | 86 |