Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003803.3 | NP_003794.3 | 23 - 24 | 3904 | 74 |
NM_019856.1 | NP_062830.1 | 22 - 23 | 3616 | 74 | |
XM_017026062.1 | XP_016881551.1 | 23 - 24 | 3631 | 74 | |
GenBank mRNA | AJ621424.1 | - | 23 - 24 | 3904 | 74 |
AK125322.1 | - | 24 - 25 | 3904 | 74 | |
BC116183.1 | - | 22 - 23 | 3357 | 74 | |
X69090.1 | - | 20 - 21 | 2985 | 74 |