Assay Design



Gene Transcripts

Gene Symbol
MYOM1
Entrez Gene ID
8736
Gene Name
myomesin 1
Gene Aliases
SKELEMIN
Chromosome Location
Chr.18: 3066807 - 3220108 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003803.3 NP_003794.3 33 - 34 4822 107
NM_019856.1 NP_062830.1 32 - 33 4534 107
XM_017026062.1 XP_016881551.1 33 - 34 4549 107
GenBank mRNA AJ621424.1 - 33 - 34 4822 107
AK125322.1 - 34 - 35 4822 107
BC116183.1 - 32 - 33 4275 107
X69090.1 - 30 - 31 3903 107

View Full Product Details