Assay Design



Gene Transcripts

Gene Symbol
MYOM1
Entrez Gene ID
8736
Gene Name
myomesin 1
Gene Aliases
SKELEMIN
Chromosome Location
Chr.18: 3066807 - 3220108 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003803.3 NP_003794.3 36 - 37 5036 98
NM_019856.1 NP_062830.1 35 - 36 4748 98
XM_017026062.1 XP_016881551.1 36 - 37 4763 98
GenBank mRNA AJ621424.1 - 36 - 37 5036 98
AK125322.1 - 37 - 38 5036 98
BC116183.1 - 35 - 36 4489 98
X69090.1 - 33 - 34 4117 98

View Full Product Details