Assay Design



Gene Transcripts

Gene Symbol
CATSPERG
Entrez Gene ID
57828
Gene Name
cation channel sperm associated auxiliary subunit gamma
Gene Aliases
C19orf15
Chromosome Location
Chr.19: 38335563 - 38370949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021185.4 NP_067008.3 26 - 27 3105 133
XM_005259114.4 XP_005259171.1 26 - 27 3201 133
XM_005259115.4 XP_005259172.1 26 - 27 3124 133
XM_005259116.2 XP_005259173.1 24 - 25 2925 133
XM_006723308.3 XP_006723371.1 26 - 27 3721 133
XM_006723310.2 XP_006723373.1 26 - 27 2988 133
XM_006723311.2 XP_006723374.1 26 - 27 3126 133
XM_011527174.2 XP_011525476.1 26 - 27 3115 133
XM_011527175.2 XP_011525477.1 26 - 27 3021 133
XM_011527176.2 XP_011525478.1 26 - 27 3024 133
XM_011527177.2 XP_011525479.1 26 - 27 2985 133
XM_011527178.2 XP_011525480.1 27 - 28 3467 133
XM_011527179.1 XP_011525481.1 26 - 27 3105 133
XM_011527182.2 XP_011525484.1 25 - 26 2849 133
XM_017027053.1 XP_016882542.1 25 - 26 2985 133
XM_017027054.1 XP_016882543.1 24 - 25 2718 133
XM_017027055.1 XP_016882544.1 13 - 14 2194 133
GenBank mRNA AK096218.1 - 1 - 2 1263 133
AK128039.1 - 2 - 3 3159 133
AK128220.1 - 26 - 27 2888 133
BC109035.1 - 22 - 23 2504 133

View Full Product Details