Assay Design



Gene Transcripts

Gene Symbol
LRRFIP2
Entrez Gene ID
9209
Gene Name
LRR binding FLII interacting protein 2
Gene Aliases
HUFI-2
Chromosome Location
Chr.3: 37052626 - 37176817 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134369.2 NP_001127841.1 2 - 3 371 197
NM_017724.2 NP_060194.1 2 - 3 371 197
XM_005265550.1 XP_005265607.1 2 - 3 338 197
XM_005265551.1 XP_005265608.1 2 - 3 338 197
XM_005265553.1 XP_005265610.1 2 - 3 338 197
XM_005265556.1 XP_005265613.1 2 - 3 338 197
XM_006713385.1 XP_006713448.1 2 - 3 338 197
XM_006713386.1 XP_006713449.1 2 - 3 338 197
XM_006713387.1 XP_006713450.1 2 - 3 338 197
XM_006713388.1 XP_006713451.1 2 - 3 338 197
XM_006713389.1 XP_006713452.1 2 - 3 338 197
XM_006713392.1 XP_006713455.1 2 - 3 338 197
XM_006713393.1 XP_006713456.1 2 - 3 338 197
XM_006713394.1 XP_006713457.1 2 - 3 338 197
XM_006713395.1 XP_006713458.1 2 - 3 338 197
XM_006713396.1 XP_006713459.1 2 - 3 338 197
XM_006713397.1 XP_006713460.1 2 - 3 338 197
XM_006713400.1 XP_006713463.1 2 - 3 338 197
XM_006713404.1 XP_006713467.1 2 - 3 338 197
XM_011534217.1 XP_011532519.1 2 - 3 338 197
XM_011534221.1 XP_011532523.1 2 - 3 338 197
XM_011534222.1 XP_011532524.1 2 - 3 338 197
XM_017007465.1 XP_016862954.1 2 - 3 338 197
XM_017007466.1 XP_016862955.1 2 - 3 338 197
XM_017007467.1 XP_016862956.1 2 - 3 338 197
XM_017007468.1 XP_016862957.1 2 - 3 338 197
XM_017007469.1 XP_016862958.1 2 - 3 338 197
XM_017007470.1 XP_016862959.1 2 - 3 338 197
XM_017007471.1 XP_016862960.1 2 - 3 338 197
XM_017007472.1 XP_016862961.1 2 - 3 338 197
XM_017007473.1 XP_016862962.1 2 - 3 338 197
XM_017007474.1 XP_016862963.1 2 - 3 338 197
XM_017007478.1 XP_016862967.1 2 - 3 338 197
XM_017007479.1 XP_016862968.1 2 - 3 338 197
XM_017007481.1 XP_016862970.1 2 - 3 338 197
XM_017007482.1 XP_016862971.1 2 - 3 338 197
GenBank mRNA AK291514.1 - 2 - 3 348 197
BC053668.1 - 2 - 3 328 197
DC336346.1 - 2 - 3 373 197

View Full Product Details