Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032493.3 | NP_115882.1 | 5 - 6 | 725 | 58 |
XM_017027421.1 | XP_016882910.1 | 5 - 6 | 665 | 58 | |
GenBank mRNA | AF290613.1 | - | 5 - 6 | 628 | 58 |
AK027528.1 | - | 5 - 6 | 633 | 58 | |
AK293188.1 | - | 4 - 5 | 509 | 58 | |
AK297824.1 | - | 5 - 6 | 668 | 58 | |
BC017469.2 | - | 5 - 6 | 670 | 58 |