Assay Design



Gene Transcripts

Gene Symbol
AP1M1
Entrez Gene ID
8907
Gene Name
adaptor related protein complex 1 mu 1 subunit
Gene Aliases
AP47, CLAPM2, CLTNM, MU-1A
Chromosome Location
Chr.19: 16197854 - 16235345 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130524.1 NP_001123996.1 8 - 9 1031 75
NM_032493.3 NP_115882.1 7 - 8 995 75
XM_017027420.1 XP_016882909.1 8 - 9 971 75
XM_017027421.1 XP_016882910.1 7 - 8 935 75
GenBank mRNA AB209808.1 - 8 - 9 951 75
AK027528.1 - 7 - 8 75
AK293188.1 - 6 - 7 779 75
AK297824.1 - 7 - 8 938 75
BC017469.2 - 7 - 8 940 75
DQ059565.1 - 8 - 9 858 75

View Full Product Details