Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161581.1 | NP_001155053.1 | 9 - 10 | 1374 | 87 |
NM_015426.4 | NP_056241.3 | 9 - 10 | 1303 | 87 | |
GenBank mRNA | AK075289.1 | - | 9 - 10 | 1018 | 87 |
AK315688.1 | - | 9 - 10 | 1025 | 87 | |
AL117629.1 | - | 4 - 5 | 342 | 87 | |
BC007417.2 | - | 8 - 9 | 953 | 87 | |
BC119692.2 | - | 9 - 10 | 1013 | 87 | |
BC119693.2 | - | 9 - 10 | 1089 | 87 |