Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001383.3 | NP_001374.3 | 9 - 10 | 76 | |
GenBank mRNA | AF321876.1 | - | 9 - 10 | 1038 | 76 |
AK090530.1 | - | 3 - 4 | 1111 | 76 | |
AK297947.1 | - | 8 - 9 | 76 | ||
AK314015.1 | - | 9 - 10 | 1024 | 76 | |
BC003099.1 | - | 9 - 10 | 1000 | 76 | |
BC096088.1 | - | 8 - 9 | 894 | 76 | |
BP417755.1 | - | 1 - 2 | 76 | ||
BT019878.1 | - | 7 - 8 | 780 | 76 | |
S81752.1 | - | 9 - 10 | 1073 | 76 | |
U34880.1 | - | 9 - 10 | 1073 | 76 |