Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164766.1 | NP_001158238.1 | 8 - 9 | 6814 | 64 |
NM_006885.3 | NP_008816.3 | 9 - 10 | 10100 | 64 | |
XM_005255957.3 | XP_005256014.1 | 9 - 10 | 18575 | 64 | |
XM_017023251.1 | XP_016878740.1 | 8 - 9 | 15810 | 64 | |
GenBank mRNA | D10250.1 | - | 8 - 9 | 6814 | 64 |
L32832.1 | - | 9 - 10 | 10100 | 64 |