Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289156.1 | NP_001276085.1 | 5 - 6 | 780 | 57 |
NM_001289157.1 | NP_001276086.1 | 5 - 6 | 725 | 57 | |
NM_002435.2 | NP_002426.1 | 6 - 7 | 908 | 57 | |
XM_011521592.1 | XP_011519894.1 | 6 - 7 | 984 | 57 | |
XM_011521593.2 | XP_011519895.1 | 6 - 7 | 966 | 57 | |
GenBank mRNA | AK225149.1 | - | 6 - 7 | 871 | 57 |
AK292374.1 | - | 6 - 7 | 861 | 57 | |
AK301370.1 | - | 5 - 6 | 749 | 57 | |
BC017351.2 | - | 5 - 6 | 674 | 57 | |
BC046357.1 | - | 6 - 7 | 863 | 57 | |
X76057.1 | - | 6 - 7 | 850 | 57 |