Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004125.2 | NP_001004125.1 | 1 - 1 | 1771 | 168 |
GenBank mRNA | AY168647.2 | - | 1 - 1 | 1353 | 168 |
BC017510.1 | - | 1 - 1 | 1146 | 168 | |
BC028316.1 | - | 1 - 1 | 870 | 168 | |
BC036931.1 | - | 1 - 1 | 849 | 168 | |
BC046919.1 | - | 1 - 1 | 1212 | 168 | |
BC110824.1 | - | 1 - 1 | 1353 | 168 |