Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318245.1 | NP_001305174.1 | 1 - 2 | 153 | 59 |
NM_032608.6 | NP_115997.5 | 1 - 2 | 153 | 59 | |
XM_011530458.2 | XP_011528760.1 | 1 - 2 | 163 | 59 | |
XM_011530461.2 | XP_011528763.1 | 1 - 2 | 163 | 59 | |
XM_011530464.2 | XP_011528766.1 | 1 - 2 | 163 | 59 | |
XM_011530466.2 | XP_011528768.1 | 1 - 2 | 164 | 59 | |
XM_017029012.1 | XP_016884501.1 | 1 - 2 | 163 | 59 | |
XM_017029014.1 | XP_016884503.1 | 1 - 2 | 163 | 59 | |
XM_017029015.1 | XP_016884504.1 | 1 - 2 | 163 | 59 | |
XM_017029017.1 | XP_016884506.1 | 1 - 2 | 162 | 59 | |
GenBank mRNA | AB075376.1 | - | 1 - 2 | 153 | 59 |
AJ310931.2 | - | 1 - 2 | 144 | 59 | |
AY077700.1 | - | 1 - 2 | 63 | 59 |