Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001485.2 | NP_001001485.1 | 14 - 15 | 1623 | 116 |
NM_001001486.1 | NP_001001486.1 | 13 - 14 | 1300 | 116 | |
NM_001001487.1 | NP_001001487.1 | 13 - 14 | 1300 | 116 | |
NM_001199179.1 | NP_001186108.1 | 14 - 15 | 1529 | 116 | |
NM_001199180.1 | NP_001186109.1 | 13 - 14 | 1518 | 116 | |
NM_001199181.1 | NP_001186110.1 | 13 - 14 | 1518 | 116 | |
NM_001199182.1 | NP_001186111.1 | 12 - 13 | 1401 | 116 | |
NM_001199183.1 | NP_001186112.1 | 13 - 14 | 1391 | 116 | |
NM_001199184.1 | NP_001186113.1 | 13 - 14 | 1391 | 116 | |
NM_001199185.1 | NP_001186114.1 | 13 - 14 | 1442 | 116 | |
NM_014382.3 | NP_055197.2 | 13 - 14 | 1442 | 116 | |
XM_005247354.2 | XP_005247411.1 | 13 - 14 | 1543 | 116 | |
XM_005247355.2 | XP_005247412.1 | 14 - 15 | 1670 | 116 | |
XM_005247356.2 | XP_005247413.1 | 14 - 15 | 1423 | 116 | |
XM_005247358.2 | XP_005247415.1 | 13 - 14 | 1247 | 116 | |
XM_011512686.2 | XP_011510988.1 | 4 - 5 | 352 | 116 | |
XM_017006164.1 | XP_016861653.1 | 14 - 15 | 1331 | 116 | |
GenBank mRNA | AB037768.1 | - | 14 - 15 | 1530 | 116 |
AB209265.1 | - | 2 - 3 | 1290 | 116 | |
AF189723.2 | - | 13 - 14 | 1274 | 116 | |
AF225981.1 | - | 13 - 14 | 1274 | 116 | |
AK074692.1 | - | 11 - 12 | 981 | 116 | |
AK296470.1 | - | 13 - 14 | 1445 | 116 | |
AK299945.1 | - | 12 - 13 | 1157 | 116 | |
AK304175.1 | - | 13 - 14 | 1247 | 116 | |
AK304374.1 | - | 13 - 14 | 1274 | 116 | |
AK314342.1 | - | 13 - 14 | 1304 | 116 | |
AY268374.1 | - | 13 - 14 | 1249 | 116 | |
AY268375.1 | - | 13 - 14 | 1249 | 116 | |
BC028139.1 | - | 13 - 14 | 1312 | 116 |