Assay Design
Gene Transcripts
- Gene Symbol
- ATP2C1
- Entrez Gene ID
-
27032
- Gene Name
- ATPase secretory pathway Ca2+ transporting 1
- Gene Aliases
-
ATP2C1A, BCPM, HHD, PMR1, SPCA1, hSPCA1
- Chromosome Location
-
Chr.3: 130850500 - 131016712 on Build GRCh38
View Full Product Details