Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001485.2 | NP_001001485.1 | 6 - 7 | 865 | 118 |
NM_001001486.1 | NP_001001486.1 | 5 - 6 | 542 | 118 | |
NM_001001487.1 | NP_001001487.1 | 5 - 6 | 542 | 118 | |
NM_001199179.1 | NP_001186108.1 | 6 - 7 | 771 | 118 | |
NM_001199180.1 | NP_001186109.1 | 5 - 6 | 760 | 118 | |
NM_001199181.1 | NP_001186110.1 | 5 - 6 | 760 | 118 | |
NM_001199182.1 | NP_001186111.1 | 4 - 5 | 643 | 118 | |
NM_001199183.1 | NP_001186112.1 | 5 - 6 | 633 | 118 | |
NM_001199184.1 | NP_001186113.1 | 5 - 6 | 633 | 118 | |
NM_001199185.1 | NP_001186114.1 | 5 - 6 | 684 | 118 | |
NM_014382.3 | NP_055197.2 | 5 - 6 | 684 | 118 | |
XM_005247354.2 | XP_005247411.1 | 5 - 6 | 785 | 118 | |
XM_005247355.2 | XP_005247412.1 | 6 - 7 | 912 | 118 | |
XM_005247356.2 | XP_005247413.1 | 6 - 7 | 665 | 118 | |
XM_005247358.2 | XP_005247415.1 | 5 - 6 | 489 | 118 | |
XM_017006164.1 | XP_016861653.1 | 6 - 7 | 573 | 118 | |
GenBank mRNA | AB037768.1 | - | 6 - 7 | 772 | 118 |
AF181120.1 | - | 5 - 6 | 597 | 118 | |
AF181121.1 | - | 5 - 6 | 597 | 118 | |
AF189723.2 | - | 5 - 6 | 516 | 118 | |
AF225981.1 | - | 5 - 6 | 516 | 118 | |
AK074692.1 | - | 3 - 4 | 223 | 118 | |
AK296470.1 | - | 5 - 6 | 687 | 118 | |
AK299945.1 | - | 4 - 5 | 399 | 118 | |
AK304175.1 | - | 5 - 6 | 489 | 118 | |
AK304374.1 | - | 5 - 6 | 516 | 118 | |
AK314342.1 | - | 5 - 6 | 546 | 118 | |
AY268374.1 | - | 5 - 6 | 491 | 118 | |
AY268375.1 | - | 5 - 6 | 491 | 118 | |
BC028139.1 | - | 5 - 6 | 554 | 118 |