Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001485.2 | NP_001001485.1 | 27 - 28 | 3136 | 117 |
NM_001199185.1 | NP_001186114.1 | 26 - 27 | 2955 | 117 | |
GenBank mRNA | AF181121.1 | - | 26 - 27 | 2868 | 117 |
BC028139.1 | - | 26 - 27 | 2825 | 117 |