Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014937.3 | NP_055752.1 | 12 - 13 | 1649 | 124 |
XM_006717720.3 | XP_006717783.1 | 12 - 13 | 4036 | 124 | |
XM_011539524.2 | XP_011537826.1 | 12 - 13 | 1412 | 124 | |
XM_011539525.2 | XP_011537827.1 | 13 - 14 | 4119 | 124 | |
XM_011539527.2 | XP_011537829.2 | 11 - 12 | 1414 | 124 | |
XM_011539528.2 | XP_011537830.1 | 11 - 12 | 1536 | 124 | |
XM_017015935.1 | XP_016871424.1 | 12 - 13 | 1403 | 124 | |
XM_017015936.1 | XP_016871425.1 | 15 - 16 | 2037 | 124 | |
GenBank mRNA | AB023183.1 | - | 12 - 13 | 1599 | 124 |
AL137528.1 | - | 6 - 7 | 614 | 124 | |
BC111493.1 | - | 12 - 13 | 1560 | 124 |