Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243195.1 | NP_001230124.1 | 1 - 2 | 313 | 118 |
NM_014937.3 | NP_055752.1 | 1 - 2 | 313 | 118 | |
XM_006717720.3 | XP_006717783.1 | 1 - 2 | 2700 | 118 | |
XM_011539525.2 | XP_011537827.1 | 1 - 2 | 2700 | 118 | |
XM_017015936.1 | XP_016871425.1 | 1 - 2 | 221 | 118 | |
GenBank mRNA | AB023183.1 | - | 1 - 2 | 263 | 118 |
AK307758.1 | - | 1 - 2 | 303 | 118 | |
BC052367.1 | - | 1 - 2 | 198 | 118 | |
BC067820.1 | - | 1 - 2 | 274 | 117 | |
BC082755.1 | - | 1 - 2 | 198 | 118 | |
BC111493.1 | - | 1 - 2 | 224 | 118 | |
DB636424.1 | - | 1 - 2 | 312 | 118 |