Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243195.1 | NP_001230124.1 | 3 - 4 | 534 | 119 |
NM_014937.3 | NP_055752.1 | 3 - 4 | 534 | 119 | |
XM_006717720.3 | XP_006717783.1 | 3 - 4 | 2921 | 119 | |
XM_011539524.2 | XP_011537826.1 | 3 - 4 | 297 | 119 | |
XM_011539525.2 | XP_011537827.1 | 4 - 5 | 3004 | 119 | |
XM_011539527.2 | XP_011537829.2 | 2 - 3 | 299 | 119 | |
XM_011539528.2 | XP_011537830.1 | 2 - 3 | 421 | 119 | |
XM_017015935.1 | XP_016871424.1 | 3 - 4 | 288 | 119 | |
XM_017015936.1 | XP_016871425.1 | 6 - 7 | 922 | 119 | |
GenBank mRNA | AB023183.1 | - | 3 - 4 | 484 | 119 |
AK307758.1 | - | 3 - 4 | 524 | 119 | |
BC052367.1 | - | 3 - 4 | 419 | 119 | |
BC067820.1 | - | 3 - 4 | 495 | 119 | |
BC082755.1 | - | 3 - 4 | 419 | 119 | |
BC111493.1 | - | 3 - 4 | 445 | 119 |