Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014937.3 | NP_055752.1 | 6 - 7 | 882 | 123 |
XM_006717720.3 | XP_006717783.1 | 6 - 7 | 3269 | 123 | |
XM_011539524.2 | XP_011537826.1 | 6 - 7 | 645 | 123 | |
XM_011539525.2 | XP_011537827.1 | 7 - 8 | 3352 | 123 | |
XM_011539527.2 | XP_011537829.2 | 5 - 6 | 647 | 123 | |
XM_011539528.2 | XP_011537830.1 | 5 - 6 | 769 | 123 | |
XM_017015935.1 | XP_016871424.1 | 6 - 7 | 636 | 123 | |
XM_017015936.1 | XP_016871425.1 | 9 - 10 | 1270 | 123 | |
GenBank mRNA | AB023183.1 | - | 6 - 7 | 832 | 123 |
AK307758.1 | - | 6 - 7 | 872 | 123 | |
BC067820.1 | - | 6 - 7 | 843 | 123 | |
BC111493.1 | - | 6 - 7 | 793 | 123 |