Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013330.4 | NP_037462.1 | 2 - 3 | 355 | 112 |
NM_197972.2 | NP_932076.1 | 2 - 3 | 324 | 112 | |
NR_104229.1 | 2 - 3 | 355 | 112 | ||
XM_017001099.1 | XP_016856588.1 | 2 - 3 | 341 | 112 | |
GenBank mRNA | AB209049.1 | - | 2 - 3 | 164 | 112 |
AF153191.2 | - | 2 - 3 | 194 | 112 | |
AK094513.1 | - | 2 - 3 | 307 | 112 | |
AK290701.1 | - | 2 - 3 | 189 | 112 | |
AK301917.1 | - | 2 - 3 | 338 | 112 | |
BC006983.1 | - | 2 - 3 | 186 | 112 | |
CD107958.1 | - | 2 - 3 | 383 | 112 |