Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080424.1 | NP_001073893.1 | 12 - 13 | 3941 | 85 |
XM_005256549.3 | XP_005256606.1 | 13 - 14 | 4093 | 85 | |
XM_005256550.4 | XP_005256607.1 | 12 - 13 | 3935 | 85 | |
XM_005256551.3 | XP_005256608.1 | 12 - 13 | 3974 | 85 | |
XM_005256552.4 | XP_005256609.1 | 12 - 13 | 4947 | 85 | |
XM_006721483.3 | XP_006721546.1 | 13 - 14 | 4038 | 85 | |
XM_011523750.2 | XP_011522052.1 | 13 - 14 | 5298 | 85 | |
XM_011523752.2 | XP_011522054.1 | 12 - 13 | 3938 | 85 | |
XM_017024381.1 | XP_016879870.1 | 12 - 13 | 5384 | 85 | |
GenBank mRNA | AB002344.2 | - | 12 - 13 | 3946 | 85 |
BC009994.1 | - | 2 - 3 | 570 | 85 |