Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206846.1 | NP_001193775.1 | 22 - 23 | 5135 | 161 |
NM_018136.4 | NP_060606.3 | 23 - 24 | 9890 | 161 | |
GenBank mRNA | AF509326.1 | - | 23 - 24 | 9633 | 161 |
AK001379.1 | - | 6 - 7 | 3024 | 161 | |
AK001411.1 | - | 6 - 7 | 1417 | 161 | |
AK226178.1 | - | 22 - 23 | 5067 | 161 | |
AY367065.1 | - | 23 - 24 | 9633 | 161 | |
AY971955.1 | - | 9 - 10 | 2388 | 161 | |
AY971956.1 | - | 22 - 23 | 4878 | 161 | |
AY971957.1 | - | 9 - 10 | 3591 | 161 | |
BC034607.1 | - | 18 - 19 | 2754 | 161 | |
BC040439.1 | - | 6 - 7 | 1452 | 161 | |
BX648804.1 | - | 6 - 7 | 4744 | 161 |