Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206846.1 | NP_001193775.1 | 24 - 25 | 5485 | 121 |
NM_018136.4 | NP_060606.3 | 25 - 26 | 10240 | 121 | |
GenBank mRNA | AF509326.1 | - | 25 - 26 | 9983 | 121 |
AK001379.1 | - | 8 - 9 | 3374 | 121 | |
AK001411.1 | - | 8 - 9 | 1767 | 121 | |
AK226178.1 | - | 24 - 25 | 5417 | 121 | |
AY367065.1 | - | 25 - 26 | 9983 | 121 | |
AY971955.1 | - | 11 - 12 | 2738 | 121 | |
AY971956.1 | - | 24 - 25 | 5228 | 121 | |
AY971957.1 | - | 11 - 12 | 3941 | 121 | |
BC034607.1 | - | 20 - 21 | 3104 | 121 | |
BC040439.1 | - | 8 - 9 | 1802 | 121 | |
BX648804.1 | - | 8 - 9 | 5094 | 121 |